Learn More
Caframo™ Chuck Guard Kit
Catalog No. 145020 Shop All Caframo Products
Change view
Click to view available options
For Use With (Equipment):
Caframo™ Overhead Stirrer
Catalog No. For Use With (Equipment)
14-502-0 Caframo™ Overhead Stirrer
1 options
Catalog No. 14-502-0 Supplier Caframo™ Supplier No. CGKIT
For retrofitting Caframo Overhead Stirrers made before 2004 (14-500-311, 14-500-10, 14-500-11 and 14-500-12)
  • Kit includes adapter and chuck guard
Specifications
For Use With (Equipment) Caframo™ Overhead Stirrer
Includes Adapter and Chuck Guard
Type Chuck Guard