Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caframo™ Chuck Set
GREENER_CHOICE

Catalog No. 145005 Shop All Caframo Products
Change view
Click to view available options
For Use With (Equipment):
Caframo™ Overhead Stirrers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-500-5 Caframo™ Overhead Stirrers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-500-5 Supplier Caframo™ Supplier No. 13CSET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chuck Set to be used with Caframo™ and Fisherbrand™ Overhead Stirrers accepting impeller shafts up to 10 mm diameter.

  • Used with Caframo High Torque and Compact overhead stirrers
  • Includes 2.5 mm hex tool for removal from stirrer output shaft for cleaning or replacement
  • Through shaft capability allows for easy adjustment of impeller height
  • Compatible with Caframo chuck guards CG1
  • Accepts shafts up to 10 mm (3/8 in.) diameter
  • 420 stainless steel
  • High strength and high wear resistance

Specifications

For Use With (Equipment) Caframo™ Overhead Stirrers
Includes Through shaft
Type Chuck Key
Weight (Metric) 280 g
Material Stainless Steel
Weight (English) 0.6 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.