Learn More
Gramercy Company LLC CAGE AUTO 5X1.25 HI-TEMP
SDP
Catalog No. NC3931229
Encompass
5x1.25 Hi-Temp Blue ElastomerThreaded Wheel/Aluminum CoreStainless Steel Precision Ball Bearing Autoclavable & Cage Washable Quiet Roll
Catalog No. NC3931229 Supplier Gramercy Company LLC Supplier No. GSCTFT51
5x1.25 Hi-Temp Blue ElastomerThreaded Wheel/Aluminum CoreStainless Steel Precision Ball Bearing Autoclavable & Cage Washable Quiet Roll