Learn More
STERIS Cage-Klenz 150 Alkaline Detergent
Catalog No. 19818864 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
Catalog No. Size
19-818-864 5 gal.
1 options
Catalog No. 19-818-864 Supplier STERIS Supplier No. 1K1505
Cage-Klenz 150 Alkaline Detergent is a chlorinated alkaline detergent specially formulated to remove difficult organic residues found in research animal caging.
  • Cage-Klenz 150 Detergent is specially formulated to meet the most challenging cleaning and materials compatibility requirements found in research animal caging.
  • Cage-Klenz 150 Detergent is a liquid, phosphate-free chlorinated alkaline detergent for use in high pressure spray animal cage washing equipment. This highly alkaline, oxidative chemistry is very effective at removing stubborn organic residues.
  • Efficacy - Chlorinated alkaline formula effectively cleans animal fats, oils and other organic cage residues in both soft and hard water
  • Flexibility - Low foaming for use in automated spray washing applications
  • Cost Savings - Reduces utility consumption and cleaning time with free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Specifications
Product Type Detergent
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Size 5 gal.