Description
- Cage-Klenz 150 Detergent is specially formulated to meet the most challenging cleaning and materials compatibility requirements found in research animal caging.
- Cage-Klenz 150 Detergent is a liquid, phosphate-free chlorinated alkaline detergent for use in high pressure spray animal cage washing equipment. This highly alkaline, oxidative chemistry is very effective at removing stubborn organic residues.
- Efficacy - Chlorinated alkaline formula effectively cleans animal fats, oils and other organic cage residues in both soft and hard water
- Flexibility - Low foaming for use in automated spray washing applications
- Cost Savings - Reduces utility consumption and cleaning time with free-rinsing formulation
- Environmentally Friendly - phosphate-free formulation
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Specifications
Specifications
| Product Type | Detergent |
| Color-coded | No |
| Form | Clear Liquid |
| Scent | Unscented |
| Sterility | Non-sterile |
| Size | 300 gal. |