Learn More
STERIS Cage-Klenz 180 Alkaline Detergent
Catalog No. 19818868 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
30 gal.
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-818-868 5 gal.
19-818-866 55 gal.
19-818-867 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-818-868 Supplier STERIS Supplier No. 1K1805
Add to Cart
Add to Cart
Cage-Klenz 180 Alkaline Detergent is a liquid, phosphate-free detergent for use in high pressure spray animal cage washing equipment. It is formulated with corrosion inhibitors for enhanced materials compatibility.
  • Cage-Klenz 180 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in research animal caging.
  • Cage-Klenz 180 Detergent is a liquid, phosphate-free detergent for use in high pressure spray animal cage washing equipment. It is formulated with corrosion inhibitors for enhanced materials compatibility.
  • Efficacy - Effectively cleans animal fats, oils and other organic cage residues in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Specifications
Product Type Detergent
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Size 5 gal.