Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Cage-Klenz 220 Acid-Based Detergent

Catalog No. 17010125 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
30 gal.
300 gal.
Container Type:
Keg
Pail
Tote
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17-010-125 30 gal. Keg
17-010-124 5 gal. Pail
17-010-123 300 gal. Tote
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-010-125 Supplier STERIS™ Supplier No. 1K2202
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cage-Klenz 220 Acid-Based Detergent is a specially formulated glycolic acid detergent used for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging. pH (1%): 2.8, typical.

Cage-Klenz 220 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.

HOW CAGE-KLENZ 220 DETERGENT WORKS

Cage-Klenz 220 Detergent is a liquid phosphate-free, hydroxyacetic acid-based detergent.

WHY CAGE-KLENZ 220 DETERGENT?

  • Efficacy - Effectively cleans hard water and urine scale, and organic soils in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Physical Properties:

pH (1%): 2.8, typical

Specifications

Product Type Acid Cleaner
Color-coded No
Form Clear Liquid
pH 2.8 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Keg
Size 30 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.