Learn More
Description
Cage-Klenz 280 Detergent is an economical detergent specially formulated to remove challenging hard water and urine scales found in research animal caging.
HOW CAGE-KLENZ 280 DETERGENT WORKS
Cage-Klenz 280 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.
WHY CAGE-KLENZ 280 DETERGENT?
- Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - formulation with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Physical Properties: pH (1%): 2.25, typical
Specifications
Specifications
| Product Type | Detergent |
| Color-coded | No |
| Form | Clear Liquid |
| pH | 2.25 (Diluted) |
| Scent | Unscented |
| Sterility | Non-sterile |
| Container Type | Drum |
| Size | 30 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.