Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Cage-Klenz 280 Acid-Based Detergent

Catalog No. 19999645 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
30 gal.
Container Type:
Drum
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
19-999-645 5 gal. Pail
19-999-644 30 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-999-645 Supplier STERIS™ Supplier No. 1K2805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cage-Klenz 280 Detergent: Economical formula for hard water and urine scale removal in research animal caging. Phosphoric acid-based, materials-compatible, cost-saving, and environmentally friendly.

Cage-Klenz 280 Detergent is an economical detergent specially formulated to remove challenging hard water and urine scales found in research animal caging.

HOW CAGE-KLENZ 280 DETERGENT WORKS

Cage-Klenz 280 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.

WHY CAGE-KLENZ 280 DETERGENT?

  • Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

Physical Properties: pH (1%): 2.25, typical

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 2.25 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.