Learn More
MilliporeSigma™ Supelco™ Cages for Fused Silica Columns
Catalog No. 111004750 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
0.25 mm I.D. Columns
Catalog No. For Use With (Equipment)
11-100-4750 0.25 mm I.D. Columns
1 options
Catalog No. 11-100-4750 Supplier MilliporeSigma™ Supelco™ Supplier No. 23742
Add to Cart
Add to Cart
Reduce the possibility of damaging the protective coating on columns; design makes it easy to hang columns in most gas chromatography ovens
  • The ends of flexible fused-silica capillary tubing are intrinsically straight and therefore the column must be coiled and properly confined on a circular frame
  • This allows for the capillary column to then be safely mounted in the column oven of the gas chromatograph
  • A unique design with minimal metal-to-column contact reduces the possibility of damaging the protective coating on columns
  • Simplifies the process of hanging columns in most GC ovens
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Hanging Columns in Gas Chromatography Ovens
For Use With (Equipment) 0.25 mm I.D. Columns
Quantity 3/Pk.
Description 6 in. (15 cm)
Product Type Fused Silica Column Cages