Learn More
Tactile Robotics Ltd CageView-SmartFeeder, TR-CV-SF-MEM500, an innovative solution designed for optimal feeding schedules and stress-free rodent environments
Catalog No. NC3357257
Encompass_Preferred
The CageView-SmartFeeder is an innovative solution designed to optimize feeding schedules while creating a stress-free environment for rodents. Seamlessly integrated with the CageView App, available for free on the Microsoft Store, this device allows precise tailoring of feeding protocols to meet experimental requirements. Researchers can manage their experiments remotely using any remote desktop access technology, providing convenience and flexibility. The SmartFeeder enables the management of feeding schedules for up to four weeks, supporting both long-term feeding and fasting protocols. Its automated feeding assessment system minimizes disturbances, ensuring a stress-free environment for rodents. Programmable and adjustable food access adds further customization options. Designed for compatibility with Tecniplast EM500 for rats and the DGM rack, the CageView-SmartFeeder is an essential tool for advanced laboratory research.
Catalog No. NC3357257 Supplier Tactile Robotics Ltd Supplier No. TRCVSFMEM500
The CageView-SmartFeeder is an innovative solution designed to optimize feeding schedules while creating a stress-free environment for rodents. Seamlessly integrated with the CageView App, available for free on the Microsoft Store, this device allows precise tailoring of feeding protocols to meet experimental requirements. Researchers can manage their experiments remotely using any remote desktop access technology, providing convenience and flexibility. The SmartFeeder enables the management of feeding schedules for up to four weeks, supporting both long-term feeding and fasting protocols. Its automated feeding assessment system minimizes disturbances, ensuring a stress-free environment for rodents. Programmable and adjustable food access adds further customization options. Designed for compatibility with Tecniplast EM500 for rats and the DGM rack, the CageView-SmartFeeder is an essential tool for advanced laboratory research.