Learn More
Tactile Robotics Ltd CageView-SmartFeeder, TR-CV-SF-MGM500 an innovative solution designed for optimal feeding schedules and stress-free rodent environments.
Catalog No. NC3357256
Encompass_Preferred
The CageView-SmartFeeder is an innovative solution designed to optimize feeding schedules and create stress-free environments for rodents. Seamlessly integrating with the CageView App, this advanced system allows researchers to tailor feeding according to experimental requirements with precision. Its automated robotic mechanism ensures precise regulation of food access based on experimental schedules, while remote access technology enables researchers to manage experiments from anywhere. The SmartFeeder supports long-term feeding and fasting schedules, allowing for comprehensive planning of up to four weeks, all while maintaining a stress-free environment for rodents. With its programmable and adjustable food access mechanisms, the SmartFeeder provides unmatched versatility and ease of use. Compatible with the Tecniplast GM500 for mice and DGM rack, the CageView-SmartFeeder is the ideal choice for modern laboratory research.
Catalog No. NC3357256 Supplier Tactile Robotics Ltd Supplier No. TRCVSFMGM500
The CageView-SmartFeeder is an innovative solution designed to optimize feeding schedules and create stress-free environments for rodents. Seamlessly integrating with the CageView App, this advanced system allows researchers to tailor feeding according to experimental requirements with precision. Its automated robotic mechanism ensures precise regulation of food access based on experimental schedules, while remote access technology enables researchers to manage experiments from anywhere. The SmartFeeder supports long-term feeding and fasting schedules, allowing for comprehensive planning of up to four weeks, all while maintaining a stress-free environment for rodents. With its programmable and adjustable food access mechanisms, the SmartFeeder provides unmatched versatility and ease of use. Compatible with the Tecniplast GM500 for mice and DGM rack, the CageView-SmartFeeder is the ideal choice for modern laboratory research.