Learn More
Tactile Robotics Ltd CageView-SmartFeeder, an innovative solution designed for optimal feeding schedules and stress-free rodent environments
Catalog No. NC3357262
Encompass_Preferred
Unlock the full potential of your rodent experiments with the CageView-SmartFeeder, an innovative solution designed to optimize feeding schedules and create stress-free rodent environments. Seamlessly integrating with the CageView app, available for free at the Microsoft Store, this user-friendly platform allows precise control over feeding and fasting schedules. The automated dispenser ensures accurate food dispensing tailored to your experiment's requirements, while detailed daily feeding schedules can be programmed for up to four weeks, ensuring consistent patterns that enhance the reliability of your research outcomes. With remote access capabilities, you can manage your experiments from anywhere using any remote desktop access technology, enabling real-time adjustments to feeding schedules for maximum flexibility.
Catalog No. NC3357262 Supplier Tactile Robotics Ltd Supplier No. CAGEVIEWSMARTFEEDERGR18
Unlock the full potential of your rodent experiments with the CageView-SmartFeeder, an innovative solution designed to optimize feeding schedules and create stress-free rodent environments. Seamlessly integrating with the CageView app, available for free at the Microsoft Store, this user-friendly platform allows precise control over feeding and fasting schedules. The automated dispenser ensures accurate food dispensing tailored to your experiment's requirements, while detailed daily feeding schedules can be programmed for up to four weeks, ensuring consistent patterns that enhance the reliability of your research outcomes. With remote access capabilities, you can manage your experiments from anywhere using any remote desktop access technology, enabling real-time adjustments to feeding schedules for maximum flexibility.