Learn More
Tactile Robotics Ltd CageView-SmartFeeder, TR-CV-SF-RGR800 an innovative solution designed for optimal feeding schedules and stress-free rodent environments.
Catalog No. NC3357258
Encompass_Preferred
The CageView-SmartFeeder is an innovative solution designed to optimize feeding schedules while creating a stress-free environment for rodents. This advanced system integrates seamlessly with the CageView app, available for free at the Microsoft Store, offering a user-friendly platform to manage your experiments with ease. Featuring a precision food dispenser, it allows you to tailor feeding schedules to meet your experimental requirements. With its automated robotic mechanism, researchers can precisely regulate food access based on experimental schedules. Remote management capabilities enable convenient control of feeding schedules from anywhere using any remote desktop access technology.
Catalog No. NC3357258 Supplier Tactile Robotics Ltd Supplier No. TRCVSFRGR800
The CageView-SmartFeeder is an innovative solution designed to optimize feeding schedules while creating a stress-free environment for rodents. This advanced system integrates seamlessly with the CageView app, available for free at the Microsoft Store, offering a user-friendly platform to manage your experiments with ease. Featuring a precision food dispenser, it allows you to tailor feeding schedules to meet your experimental requirements. With its automated robotic mechanism, researchers can precisely regulate food access based on experimental schedules. Remote management capabilities enable convenient control of feeding schedules from anywhere using any remote desktop access technology.