Learn More
Tactile Robotics Ltd CageView-SmartVision, TR-CV-SV-UNIVERSAL, an innovative animal tracking device designed to elevate research efficiency and accuracy
Catalog No. NC3357259
Encompass_Preferred
Elevate your animal behavior research with CageView-SmartVision, a cutting-edge animal tracking device designed to enhance efficiency and precision. Offering 24/7 monitoring, remote access, and an AI-powered mouse detection system, this innovative solution simplifies behavioral studies while remaining cost-effective. The CageView-SmartVision seamlessly integrates with the CageView App, available for free on the Microsoft Store, enabling advanced animal tracking and meeting the demands of rigorous research protocols. Its user-friendly design supports versatile applications, enhancing research efficiency while keeping costs low. Compatible with all cages within the CageView system, SmartVision can also be used in open fields to collect data and record motion, providing comprehensive control over animal behavior for deeper insights.
Catalog No. NC3357259 Supplier Tactile Robotics Ltd Supplier No. TRCVSVUNIVERSAL
Elevate your animal behavior research with CageView-SmartVision, a cutting-edge animal tracking device designed to enhance efficiency and precision. Offering 24/7 monitoring, remote access, and an AI-powered mouse detection system, this innovative solution simplifies behavioral studies while remaining cost-effective. The CageView-SmartVision seamlessly integrates with the CageView App, available for free on the Microsoft Store, enabling advanced animal tracking and meeting the demands of rigorous research protocols. Its user-friendly design supports versatile applications, enhancing research efficiency while keeping costs low. Compatible with all cages within the CageView system, SmartVision can also be used in open fields to collect data and record motion, providing comprehensive control over animal behavior for deeper insights.