Learn More
Medchemexpress LLC Cagrilintide | 1415456-99-3 | 100.0% | 4409.01 | 5 MG
SDP
Catalog No. 5000396981
Encompass_Preferred
Cagrilintide is an investigational novel long-acting acylated amylin analogue. It acts as a nonselective amylin receptors (AMYR) and calcitonin G protein-coupled receptor (CTR) agonist. Cagrilintide induces significant weight loss and reduces food intake, showing potential for obesity research.
  • Investigational novel long-acting acylated amylin analogue
  • Acts as a nonselective amylin and calcitonin G protein-coupled receptor agonist
  • Induces significant weight loss and reduces food intake
  • Potential for obesity research
  • White to off-white solid appearance
  • Soluble in H2O at 100 mg/mL (requires ultrasonic)
  • Store powder at -80°C for 2 years or -20°C for 1 year (sealed, dry, under nitrogen)
  • Store solutions at -80°C for 6 months or -20°C for 1 month (sealed, dry, under nitrogen)
Catalog No. 50-003-96981 Supplier Medchemexpress LLC Supplier No. HYP34625MG
Cagrilintide is an investigational novel long-acting acylated amylin analogue. It acts as a nonselective amylin receptors (AMYR) and calcitonin G protein-coupled receptor (CTR) agonist. Cagrilintide induces significant weight loss and reduces food intake, showing potential for obesity research.
  • Investigational novel long-acting acylated amylin analogue
  • Acts as a nonselective amylin and calcitonin G protein-coupled receptor agonist
  • Induces significant weight loss and reduces food intake
  • Potential for obesity research
  • White to off-white solid appearance
  • Soluble in H2O at 100 mg/mL (requires ultrasonic)
  • Store powder at -80°C for 2 years or -20°C for 1 year (sealed, dry, under nitrogen)
  • Store solutions at -80°C for 6 months or -20°C for 1 month (sealed, dry, under nitrogen)