Learn More
Medchemexpress LLC Cagrilintide acetate | 1415456-99-3 | CB712152538 | 99.96% | 4409.01 | 5 MG
SDP
Catalog No. 5000397367
Encompass_Preferred
Cagrilintide acetate is a non-selective AMYR/CTR agonist and a long-acting acylated amylase analogue. It reduces food intake and causes significant weight loss in a dose-dependent manner, making it suitable for obesity studies.
  • Non-selective AMYR/CTR agonist
  • Long-acting acylated amylase analogue
  • Reduces food intake
  • Causes significant weight loss in a dose-dependent manner
  • Suitable for obesity studies
  • For research use only
Catalog No. 50-003-97367 Supplier Medchemexpress LLC Supplier No. HYP3462A5MG
Cagrilintide acetate is a non-selective AMYR/CTR agonist and a long-acting acylated amylase analogue. It reduces food intake and causes significant weight loss in a dose-dependent manner, making it suitable for obesity studies.
  • Non-selective AMYR/CTR agonist
  • Long-acting acylated amylase analogue
  • Reduces food intake
  • Causes significant weight loss in a dose-dependent manner
  • Suitable for obesity studies
  • For research use only