Learn More
System Biosciences LLC CAGs-GFP-T2A-Luciferase Enhanced Episomal Vector (EEV)CAGs-GFP-T2A-Luciferase Dual Reporter Enhanced Episomal Vector (EEV) = 10ug
SDP
Catalog No. NC2164078
Encompass_Preferred
Get sustained, non-viral constitutive expression of GFP and luciferase reporters with this EEV plasmid and avoid the challenges of viral transduction. With virtually no limits on insert size (unlike AAV vectors) Enhanced Episomal Vectors (EEVs) are an excellent choice for non-integrating, non-viral gene expression. Because they replicate in synchrony with the host cell, they are stably inherited and can be used for long-lasting expression—up to several months both in vitro and in vivo—without modifying the host genome. Use SBI’s CAGs-GFP-T2A-Luciferase EEV Vector for constitutive, CAG-driven coordinated expression of GFP and luciferase reporters in most cell types, including primary cells and stem cells.
This item is not returnable. View return policy
Catalog No. NC2164078 Supplier System Biosciences LLC Supplier No. EEV604A2
Get sustained, non-viral constitutive expression of GFP and luciferase reporters with this EEV plasmid and avoid the challenges of viral transduction. With virtually no limits on insert size (unlike AAV vectors) Enhanced Episomal Vectors (EEVs) are an excellent choice for non-integrating, non-viral gene expression. Because they replicate in synchrony with the host cell, they are stably inherited and can be used for long-lasting expression—up to several months both in vitro and in vivo—without modifying the host genome. Use SBI’s CAGs-GFP-T2A-Luciferase EEV Vector for constitutive, CAG-driven coordinated expression of GFP and luciferase reporters in most cell types, including primary cells and stem cells.