Learn More
Thomas Scientific CAHN MICROBALANCE PANS"A"100PK
SDP
Catalog No. NC1663200
Encompass
Cahn Microbalance Pans, Style A, 12mm diameter, 17mg each
Catalog No. NC1663200 Supplier Thomas Scientific Supplier No. 0249A56
Cahn Microbalance Pans, Style A, 12mm diameter, 17mg each