Learn More
MSC PIP Welding Glove, Elk Grain Leather, Insulated, Kevlar Stitched, 1/Pr.
Catalog No. 5000688934
Encompass
These premium welding gloves are designed for MIG and stick welding, offering a combination of comfort, protection, and durability. Constructed with high-grade elkskin leather, the gloves feature 3-dimensional pre-curved fingers for a form-fitting design that improves dexterity.
  • Constructed from premium gold and gray elkskin leather with completely welted seams
  • Features Kevlar® thread stitching for increased heat and flame resistance
  • Equipped with a flame-resistant wool-insulated back and pulse protector for added safety
  • Includes a reinforced palm and thumb strap for enhanced durability and comfort
  • Designed with an extended gauntlet cuff for additional wrist protection
  • Includes a 3 in. carabiner for convenient storage and organization
Catalog No. 50-006-88934 Supplier MSC Supplier No. 19907930
These premium welding gloves are designed for MIG and stick welding, offering a combination of comfort, protection, and durability. Constructed with high-grade elkskin leather, the gloves feature 3-dimensional pre-curved fingers for a form-fitting design that improves dexterity.
  • Constructed from premium gold and gray elkskin leather with completely welted seams
  • Features Kevlar® thread stitching for increased heat and flame resistance
  • Equipped with a flame-resistant wool-insulated back and pulse protector for added safety
  • Includes a reinforced palm and thumb strap for enhanced durability and comfort
  • Designed with an extended gauntlet cuff for additional wrist protection
  • Includes a 3 in. carabiner for convenient storage and organization