Learn More
Med Vet International Caiman Forceps 5mm X 240mm, J1480A, Each
Catalog No. 5000585780
Encompass_Preferred
Caiman is intended to be used as a multipurpose vessel sealing and cutting instrument in laparoscopic, thorascopic, and open surgery. The Caiman uses a radio frequency generator connected via an electric cord to an all-in-one instrument that will safely seal, cauterize and cut soft tissue. Plug and Play Easy and safe to use Long jaw tip Enlarged vessel sealing length and improved surgical efficiency Tip first closure Retains tissues within the jaws for improved compression Uniform tissue compression Leads to consistent sealing quality from distal to proximal tip One seal confidence State of the art vessel sealing with only one step energy activation Caiman instruments seal vessels and vessel bundles up to 7mm in diameter and feature an average thermal spread <1mm. Effectively seals with virtually no adhesion or charring. The Caiman takes care of temperature, pressure and time of sealing.
Catalog No. 50-005-85780 Supplier Med Vet International Supplier No. JOR1480A
Caiman is intended to be used as a multipurpose vessel sealing and cutting instrument in laparoscopic, thorascopic, and open surgery. The Caiman uses a radio frequency generator connected via an electric cord to an all-in-one instrument that will safely seal, cauterize and cut soft tissue. Plug and Play Easy and safe to use Long jaw tip Enlarged vessel sealing length and improved surgical efficiency Tip first closure Retains tissues within the jaws for improved compression Uniform tissue compression Leads to consistent sealing quality from distal to proximal tip One seal confidence State of the art vessel sealing with only one step energy activation Caiman instruments seal vessels and vessel bundles up to 7mm in diameter and feature an average thermal spread <1mm. Effectively seals with virtually no adhesion or charring. The Caiman takes care of temperature, pressure and time of sealing.