Learn More
Cayman Chemical CAIX InhIbItr S4 10mg
SDP
Catalog No. 502989406
Encompass_Preferred
A CAIX inhibitor (Ki = 0.007 µM for the recombinant human enzyme); selective for CAIX over CAI and CAII but does inhibit CAXII (Kis = 5.6, 0.55, and 0.002 µM, respectively); inhibits proliferation of HT-29 and MDA-MB-231, but not HCT116, cancer cells under anoxic conditions (IC50s = 20, 481, and >1,000 µM, respectively); reduces the number of lung metastases, but not primary tumor growth, in an MDA-MB-231 mouse xenograft model at 10 mg/kg
Catalog No. 50-298-9406 Supplier Cayman Chemical Supplier No. 3111910 MG
Add to Cart
Add to Cart
A CAIX inhibitor (Ki = 0.007 µM for the recombinant human enzyme); selective for CAIX over CAI and CAII but does inhibit CAXII (Kis = 5.6, 0.55, and 0.002 µM, respectively); inhibits proliferation of HT-29 and MDA-MB-231, but not HCT116, cancer cells under anoxic conditions (IC50s = 20, 481, and >1,000 µM, respectively); reduces the number of lung metastases, but not primary tumor growth, in an MDA-MB-231 mouse xenograft model at 10 mg/kg