Learn More
Foxx Life Sciences Autofil Sterile Disposable Centrifuge Tube Vacuum Filter Units with 0.2um PES Membrane, 15mL, 24/CS
Supplier:  Foxx Life Sciences 1411RLS
Encompass_Preferred
1411-RLS: Full Assembly, 15mL, 0.2um PES. Autofil is available as a complete system, or as a funnel-only bottle-top vacuum filter. The patented, next-generation product comes with 3 different asymmetric PES membrane pore sizes (0.1um, 0.2um, or 0.45um). Our membrane is low protein-binding and surfactant-free. The Autofil system is designed for sterilization or clarification of aqueous laboratory fluids. The filters are sterilized in the USA. An industry first, Foxx Life Sciences offers a stable, low-profile, patented optional pedestal base or low-cost ring stand cradle. Each holder provides a stationary vacuum hose connection that makes setup and use quick and efficient, while preventing spills and tissue culture hood contamination. These holders also allow for hands-free operation of the filter unit. Foxx Life Sciences' Autofil is manufactured in an ISO 13485 clean room in NA.
Catalog No. 50-104-9880