Learn More
Foxx Life Sciences EZFlow Hydrophobic PTFE Membrane Filters, 0.45um, 90mm Diameter Filter Discs, 25/PK
Catalog No. 501049909
Encompass_Preferred
365-3811-OEM: EZFlow Membrane Disc Filter, 0.45um Hydrophobic PTFE, 90mm, 25/pack. EZFlow 13mm, 25mm, 47mm, and 90mm membrane disc filters are used to retain particles or microorganisms by surface capture. The membrane disc filters are available in Nylon or Hydrophobic PTFE with polypropylene support, in 0.2um or 0.45um pore sizes. The Nylon membrane has great chemical resistance, and is ideal for filtering more aggressive solutions such as alcohols and DMSO. The Hydrophobic PTFE membrane is bonded to a polypropylene support. Due to its extreme chemical compatibility, they are ideal for the filtration of solvents and acids, or other aggressive chemicals. All materials comply with USP Class VI Standards and FDA compliances.
Catalog No. 50-104-9909 Supplier Foxx Life Sciences Supplier No. 3653811OEM
365-3811-OEM: EZFlow Membrane Disc Filter, 0.45um Hydrophobic PTFE, 90mm, 25/pack. EZFlow 13mm, 25mm, 47mm, and 90mm membrane disc filters are used to retain particles or microorganisms by surface capture. The membrane disc filters are available in Nylon or Hydrophobic PTFE with polypropylene support, in 0.2um or 0.45um pore sizes. The Nylon membrane has great chemical resistance, and is ideal for filtering more aggressive solutions such as alcohols and DMSO. The Hydrophobic PTFE membrane is bonded to a polypropylene support. Due to its extreme chemical compatibility, they are ideal for the filtration of solvents and acids, or other aggressive chemicals. All materials comply with USP Class VI Standards and FDA compliances.