Learn More
Foxx Life Sciences EZBio Gridded Sterile Mixed Cellulose Ester (MCE) Membrane Filters, 0.45um, White, 100/Pack
Supplier:  Foxx Life Sciences 36E3613OEM
Encompass_Preferred
36E-3613-OEM: EZBio Gridded Disc Filter, 0.45um MCE, White and Gridded, 47mm, Sterile, 100/pack. EZBio 47mm gridded disc filters are used for microbiology analysis of water and other liquids in the food, beverage, and pharmaceutical industries. The material of the gridded membrane is mixed cellulose ester with a 0.45um pore size. Each gridded disc filter is sterilized by gamma irradiation and individually packaged. All materials comply with USP Class VI Standards and FDA compliances.
Catalog No. 50-104-9910