Learn More
Foxx Life Sciences EZFlow Sterile Hydrophilic PVDF Syringe Filters, 0.22um Sterilizing Membrane, 25mm, 100/PK
Supplier:  Foxx Life Sciences 3782215OEM
Encompass_Preferred
378-2215-OEM: EZFlow Syringe Filter, 0.22um Hydrophilic PVDF, 25mm, Sterile, 100/pack. EZFlow 13mm, 25mm, and 33mm Syringe Filters are used for prefiltration, clarification, sterilization, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials in both 0.2um and 0.45um pore sizes. The filter membranes are reinforced with polypropylene housing, with a female Luer-lok inlet and a male Luer slip outlet. All materials comply with USP Class VI Standards and are available for both sterile and non-sterile applications.
Catalog No. 50-104-9920