Learn More
ATCC Caki-1; Kidney Carcinoma; Human (Homo sapiens)
Catalog No. 502384482
Encompass
Caki-1 is a cell line with epithelial morphology that was isolated from kidney tissue obtained from a 49-year-old, White, male patient with clear cell carcinoma. This cell line is a suitable transfection host and has applications in cancer and toxicology research.
Catalog No. 50-238-4482 Supplier ATCC Supplier No. HTB46
Caki-1 is a cell line with epithelial morphology that was isolated from kidney tissue obtained from a 49-year-old, White, male patient with clear cell carcinoma. This cell line is a suitable transfection host and has applications in cancer and toxicology research.