Learn More
Selleck Chemical LLC Idelalisib (CAL-101, GS-1101) 10mg 870281-82-6
Supplier: Selleck Chemical LLC S222610MG
Idelalisib (CAL-101, GS-1101) is a selective p110? inhibitor with IC50 of 2.5 nM in cell-free assays; shown to have 40- to 300-fold greater selectivity for p110? than p110?/?/?, and 400- to 4000-fold more selectivity to p110? than C2?, hVPS34, DNA-PK and mTOR. Idelalisib also stimulates autophagy. *For Research & Development use only. Product is not intended for drug, household, or human consumption.