Learn More
Selleck Chemical LLC Idelalisib (CAL-101, GS-1101) 50mg 870281-82-6
SDP
Catalog No. 508608
Encompass_Preferred
Idelalisib (CAL-101, GS-1101) is a selective p110? inhibitor with IC50 of 2.5 nM in cell-free assays; shown to have 40- to 300-fold greater selectivity for p110? than p110?/?/?, and 400- to 4000-fold more selectivity to p110? than C2?, hVPS34, DNA-PK and mTOR. Idelalisib also stimulates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-860-8 Supplier Selleck Chemical LLC Supplier No. S222650MG
Add to Cart
Add to Cart
Idelalisib (CAL-101, GS-1101) is a selective p110? inhibitor with IC50 of 2.5 nM in cell-free assays; shown to have 40- to 300-fold greater selectivity for p110? than p110?/?/?, and 400- to 4000-fold more selectivity to p110? than C2?, hVPS34, DNA-PK and mTOR. Idelalisib also stimulates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.