Learn More
Apexbio Technology LLC CAL-101 (Idelalisib, GS-1101), 5mg. Cas: 870281-82-6 MFCD: MFCD19443647. PI3K inhibitor
SDP
Supplier:  Apexbio Technology LLC A30055
Encompass_Preferred
CAL-101 (Idelalisib, GS-1101), is a p110 selective phosphatidylinositol-3-kinase inhibitor in a kinome-wide screen using purified enzymes and in cell-based PI3K isoform-specific assays. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4776