Learn More
Medchemexpress LLC Cal-130 hydrochloride | 1431697-78-7 | 98.7% | 462.93 g/mol | C23H23ClN8O | 10 MG
SDP
Catalog No. 5000288600
Encompass_Preferred
Cal-130 hydrochloride is the hydrochloride salt of a small-molecule inhibitor selective for PI3Kδ (p110δ) and PI3Kγ (p110γ). It is supplied as a solid research reagent and shows low-nanomolar potency against its primary targets, with weaker activity toward other PI3K isoforms.
  • Selective inhibition of PI3Kδ and PI3Kγ, low-nanomolar IC50 values
  • Demonstrates activity against additional PI3K isoforms at higher concentrations
  • High chemical purity suitable for in vitro and preclinical research
  • Supplied as a solid, with common pack sizes suitable for laboratory studies
  • Provided with documentation that includes molecular formula, molecular weight, and CAS number
Catalog No. 50-002-88600 Supplier Medchemexpress LLC Supplier No. HY16122B10MG
Cal-130 hydrochloride is the hydrochloride salt of a small-molecule inhibitor selective for PI3Kδ (p110δ) and PI3Kγ (p110γ). It is supplied as a solid research reagent and shows low-nanomolar potency against its primary targets, with weaker activity toward other PI3K isoforms.
  • Selective inhibition of PI3Kδ and PI3Kγ, low-nanomolar IC50 values
  • Demonstrates activity against additional PI3K isoforms at higher concentrations
  • High chemical purity suitable for in vitro and preclinical research
  • Supplied as a solid, with common pack sizes suitable for laboratory studies
  • Provided with documentation that includes molecular formula, molecular weight, and CAS number