Learn More
Radiometer America Radiometer Gas CAL 2 for ABL800 FLEX Blood Gas Analyzer
SDP
Catalog No. NC1065975
Encompass_Preferred
ABL800 FLEX CO2 Gas CAL 2 (10 bar)
Calibration gas for the ABL800 FLEX blood gas analyzer. 
ABL800 FLEX Blood Gas Analyzer
With 15,000 active analyzers globally, the ABL800 FLEX analyzer provides you with proven performance. The accuracy and precision of the analyzer are based on advanced sensor technology, an optical measuring system and the fact that several of the steps normally associated with blood gas testing are fully automated when using the ABL800 FLEX analyzer. As a result, you get high analytical performance in the measurement of pH, blood gases, electrolytes, metabolites and oximetry parameters.
Catalog No. NC1065975 Supplier Radiometer America Supplier No. 962175
ABL800 FLEX CO2 Gas CAL 2 (10 bar)
Calibration gas for the ABL800 FLEX blood gas analyzer. 
ABL800 FLEX Blood Gas Analyzer
With 15,000 active analyzers globally, the ABL800 FLEX analyzer provides you with proven performance. The accuracy and precision of the analyzer are based on advanced sensor technology, an optical measuring system and the fact that several of the steps normally associated with blood gas testing are fully automated when using the ABL800 FLEX analyzer. As a result, you get high analytical performance in the measurement of pH, blood gases, electrolytes, metabolites and oximetry parameters.