Learn More
ATCC CAL 27; Tongue Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2095
Encompass
Cal 27 cells are epithelial cells isolated from tissue taken prior to treatment from a 56-year-old, White male with a lesion in the middle of the tongue in 1982.
Catalog No. 50-238-1686