Learn More
Aat Bioquest Cal-520, AM | 10x50ug
SDP
Catalog No. NC0831165
Encompass_Preferred
Cal-520 AM provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization. Cal-520 AM is a new fluorogenic calcium-sensitive dye with a significantly improved signal to noise ratio and intracellular retention compared to the existing green calcium indicators (such as Fluo-3 AM and Fluo-4 AM). Cells expressing a GPCR or calcium channel of interest that signals through calcium can be preloaded with Cal-520 AM which can cross cell membrane. Once inside the cell, the lipophilic blocking groups of Cal-520™ AM are cleaved by esterases, resulting in a negatively charged fluorescent dye that stays inside cells. Its fluorescence is greatly enhanced upon binding to calcium. When cells stimulated with agonists, the receptor signals the release of intracellular calcium, which significantly increase the fluorescence of Cal-520.
Catalog No. NC0831165 Supplier Aat Bioquest Supplier No. 21130
Add to Cart
Edge
Add to Cart
Cal-520 AM provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization. Cal-520 AM is a new fluorogenic calcium-sensitive dye with a significantly improved signal to noise ratio and intracellular retention compared to the existing green calcium indicators (such as Fluo-3 AM and Fluo-4 AM). Cells expressing a GPCR or calcium channel of interest that signals through calcium can be preloaded with Cal-520 AM which can cross cell membrane. Once inside the cell, the lipophilic blocking groups of Cal-520™ AM are cleaved by esterases, resulting in a negatively charged fluorescent dye that stays inside cells. Its fluorescence is greatly enhanced upon binding to calcium. When cells stimulated with agonists, the receptor signals the release of intracellular calcium, which significantly increase the fluorescence of Cal-520.