Learn More
Abcam Cal-520, AM
Catalog No. NC3863889
Encompass_Preferred
Cal-520, AM is a fluorogenic calcium-sensitive dye for detecting intracellular calcium mobilization with significantly improved signal-to-noise ratio and intracellular retention compared to existing calcium indicators.
Catalog No. NC3863889 Supplier Abcam Supplier No. AB171868
Add to Cart
Add to Cart
Cal-520, AM is a fluorogenic calcium-sensitive dye for detecting intracellular calcium mobilization with significantly improved signal-to-noise ratio and intracellular retention compared to existing calcium indicators.