Learn More
Grainger Cal Gas 116L Hydrogen Cyanide Nitrogen
Catalog No. 502767724
Encompass_Preferred
The GASCO Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 103L cylinder with 99.998% nitrogen. The cylinder has a CGA 580 outlet connection and is rated for service pressure up to 3000 psig. This calibration gas meets standards for instrument calibration gas and provides accurate, repeatable results when used for calibration and bump testing. The cylinder is refillable and is a reliable nitrogen gas source for ongoing calibration needs.
Catalog No. 50-276-7724 Supplier Grainger Supplier No. 16N145
The GASCO Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 103L cylinder with 99.998% nitrogen. The cylinder has a CGA 580 outlet connection and is rated for service pressure up to 3000 psig. This calibration gas meets standards for instrument calibration gas and provides accurate, repeatable results when used for calibration and bump testing. The cylinder is refillable and is a reliable nitrogen gas source for ongoing calibration needs.