Learn More
Grainger Cal Gas 116L Hydrogen Cyanide Nitrogen
Catalog No. 502766163
Encompass_Preferred
The Grainger 16N144 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications and is ideal for pumps, fans, and conveyors. The motor is CSA certified and backed by a 1 year warranty from Grainger.
Catalog No. 50-276-6163 Supplier Grainger Supplier No. 16N144
The Grainger 16N144 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications and is ideal for pumps, fans, and conveyors. The motor is CSA certified and backed by a 1 year warranty from Grainger.