Learn More
Grainger Cal Gas 116L Hydrogen Sulfide Nitrogen
Catalog No. 502765334
Encompass_Preferred
The Grainger 16N158 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a C-face mounting style. The 16N158 is designed for general purpose applications.
Catalog No. 50-276-5334 Supplier Grainger Supplier No. 16N158
The Grainger 16N158 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a C-face mounting style. The 16N158 is designed for general purpose applications.