Learn More
Grainger Cal Gas 116L Hydrogen Sulfide Air
Catalog No. 502767657
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing zero air. It has a capacity of 103L and is filled with 20.9% oxygen and 79.1% nitrogen. This high pressure cylinder has a CGA 590 outlet connection and is used for calibration and bump testing of gas detection instruments. The cylinder is manufactured by GASCO in the United States and is certified to meet NIST traceable standards. The cylinder must be secured upright during storage and transport. Proper safety precautions should be followed when handling compressed gas cylinders. This calibration gas is suitable for various industrial, safety, and scientific applications that require zero air reference gas.
Catalog No. 50-276-7657 Supplier Grainger Supplier No. 16N156
The GASCO Calibration Gas - Air is a calibration gas cylinder containing zero air. It has a capacity of 103L and is filled with 20.9% oxygen and 79.1% nitrogen. This high pressure cylinder has a CGA 590 outlet connection and is used for calibration and bump testing of gas detection instruments. The cylinder is manufactured by GASCO in the United States and is certified to meet NIST traceable standards. The cylinder must be secured upright during storage and transport. Proper safety precautions should be followed when handling compressed gas cylinders. This calibration gas is suitable for various industrial, safety, and scientific applications that require zero air reference gas.