Learn More
Grainger Cal Gas 116L Hydrogen Sulfide Air
Catalog No. 502768316
Encompass_Preferred
The Grainger 16N152 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It comes with standard features like ball bearings, cast iron end brackets, and a cast iron conduit box. This ODP motor would be well suited for driving pumps, fans, conveyors, compressors, and other industrial equipment. With proper installation and maintenance, this motor should provide years of reliable operation.
Catalog No. 50-276-8316 Supplier Grainger Supplier No. 16N152
The Grainger 16N152 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It comes with standard features like ball bearings, cast iron end brackets, and a cast iron conduit box. This ODP motor would be well suited for driving pumps, fans, conveyors, compressors, and other industrial equipment. With proper installation and maintenance, this motor should provide years of reliable operation.