Learn More
Grainger Gasco Calibration Gas, Nitrogen Dioxide in Air, 10 ppm, 116 L
Catalog No. 502768368
Encompass_Preferred
This calibration gas is designed for the accurate testing and maintenance of gas detection equipment. Formulated with 10 ppm nitrogen dioxide in an air balance, it ensures reliable performance and precise sensor response verification. The aluminum alloy cylinder provides a durable, lightweight container for this 116 L gas supply, suitable for a wide range of analytical applications.
  • Contains 10 ppm nitrogen dioxide in air
  • 116 L cylinder capacity
  • ±5% accuracy for dependable results
  • Features a C-10 connection for wide compatibility
  • Includes NIST calibration certificate
  • Constructed from aluminum alloy
  • Compatible with all OEM gas detectors
Catalog No. 50-276-8368 Supplier Grainger Supplier No. 16N167
This calibration gas is designed for the accurate testing and maintenance of gas detection equipment. Formulated with 10 ppm nitrogen dioxide in an air balance, it ensures reliable performance and precise sensor response verification. The aluminum alloy cylinder provides a durable, lightweight container for this 116 L gas supply, suitable for a wide range of analytical applications.
  • Contains 10 ppm nitrogen dioxide in air
  • 116 L cylinder capacity
  • ±5% accuracy for dependable results
  • Features a C-10 connection for wide compatibility
  • Includes NIST calibration certificate
  • Constructed from aluminum alloy
  • Compatible with all OEM gas detectors