Learn More
Grainger Cal Gas 116L Sulfur Dioxide Nitrogen
Catalog No. 502766790
Encompass_Preferred
The Grainger 16N177 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This 10 lb stainless steel cylinder extinguisher uses Novec 1230 fluid which leaves no residue and is safe for use around sensitive electronics. It has a discharge range of 10-15 ft and can operate between -4°F to 120°F. The extinguisher meets UL and ULC standards and comes with a 5 year limited warranty from the manufacturer.
Catalog No. 50-276-6790 Supplier Grainger Supplier No. 16N177
The Grainger 16N177 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This 10 lb stainless steel cylinder extinguisher uses Novec 1230 fluid which leaves no residue and is safe for use around sensitive electronics. It has a discharge range of 10-15 ft and can operate between -4°F to 120°F. The extinguisher meets UL and ULC standards and comes with a 5 year limited warranty from the manufacturer.