Learn More
Gasco Chlorine 10 PPM in Nitrogen Calibration Gas 58 Liter Cylinder C-10 Connection
Catalog No. NC1836974
Encompass_Preferred
Chlorine 10 PPM in Nitrogen Calibration Gas contained in a 58 Liter Aluminum Cylinder, Cylinder connection C-10 5/8"-18 UNF, Calibration Certificate NIST, Made in the USA.
Catalog No. NC1836974 Supplier Gasco Supplier No. 58L25210
Chlorine 10 PPM in Nitrogen Calibration Gas contained in a 58 Liter Aluminum Cylinder, Cylinder connection C-10 5/8"-18 UNF, Calibration Certificate NIST, Made in the USA.