Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gasco Isobutylene | 115-11-7 | MFCD00008898 | 103L

Catalog No. NC1575895
Encompass_Preferred

Isobutylene | 100ppm | MW: 56.107 | 115-11-7 | MFCD00008898 | 103L

Catalog No. NC1575895 Supplier Gasco Supplier No. 103L248100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Isobutylene | 100ppm | MW: 56.107 | 115-11-7 | MFCD00008898 | 103L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.