Learn More
Aat Bioquest Cal Red R525/650 AM (1 mg)
SDP
Catalog No. 502249869
Encompass_Preferred
The intracellular calcium flux assay is a widely used method for monitoring the activities of GPCRs and calcium channels. This catalog is for 1 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9869 Supplier Aat Bioquest Supplier No. 20590
The intracellular calcium flux assay is a widely used method for monitoring the activities of GPCRs and calcium channels. This catalog is for 1 mg.