Learn More
Pickering Laboratories Inc CAL STANDARD 5ML
SDP
Catalog No. NC9230843
Encompass_Preferred
Native Sample Standard without Norleucine & Alpha-Amino-Beta-Guanidinopropionic Acid in Lithium Citrate Buffer, 0.25 µmole/mL, each (5 mL /bottle) Amino Acid Analysis.
Catalog No. NC9230843 Supplier Pickering Laboratories Inc Supplier No. 17000170
Native Sample Standard without Norleucine & Alpha-Amino-Beta-Guanidinopropionic Acid in Lithium Citrate Buffer, 0.25 µmole/mL, each (5 mL /bottle) Amino Acid Analysis.