Learn More
Perkin Elmer US LLC 12-Element Solution, H2O/trace HF/trace HNO3, 10 µg/mL, 125mL
Catalog No. 509045753
Encompass_Preferred
Details Concentration Element 10 µg/mL B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr
Catalog No. 50-904-5753 Supplier Perkin Elmer US LLC Supplier No. N9300235
Details Concentration Element 10 µg/mL B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr