Learn More
RNA Medical CAL VERIFY BLOOD GAS CTRL
SDP
Catalog No. NC9980297
Encompass
RNA; RNA Medical Brand; CVC 123; Verify the performance, linearity of Analysers that measure PH, Blood Gases, electrolytes, glucose lactate, document proper calibration, confirm preventive maintenence; Assist w/regulatory complian
Catalog No. NC9980297 Supplier RNA Medical Supplier No. CVC 123
RNA; RNA Medical Brand; CVC 123; Verify the performance, linearity of Analysers that measure PH, Blood Gases, electrolytes, glucose lactate, document proper calibration, confirm preventive maintenence; Assist w/regulatory complian