Learn More
RNA Medical CO-Oximeter Calibration Verification Controls
SDP
Catalog No. nc9980298
Encompass
Kit, Control; CO-Oximeter Calibration Verification; Multi-Level CO-Oximeter; (20 amps X 1.2mL, 4 amps each level); CVC 223 has a shelf life of eighteen (18) months from the date of manufacture when stored at 2 deg. to 8 deg.C
Catalog No. NC9980298 Supplier RNA Medical Supplier No. CVC 223
Kit, Control; CO-Oximeter Calibration Verification; Multi-Level CO-Oximeter; (20 amps X 1.2mL, 4 amps each level); CVC 223 has a shelf life of eighteen (18) months from the date of manufacture when stored at 2 deg. to 8 deg.C