Learn More
Aviva Systems Biology Corp CALB1 Antibody - C-terminal region (ARP60104_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60104_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CALB1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CALB1. Alternate names: CALBApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-943